missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethyl-2,5-hexanediol, 97%
Description
It is used as an intermediate in the preparation of 2.5-Dimethyl-2.5-bis(tertbutyl-peroxy)hexane to produce polyethylene copolymers and polyethylene rubbers. It is also used in the synthesis of six- and seven-membered heterocyclic boron compounds containing intramolecular N-B bond.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an intermediate in the preparation of 2.5-Dimethyl-2.5-bis(tertbutyl-peroxy)hexane to produce polyethylene copolymers and polyethylene rubbers. It is also used in the synthesis of six- and seven-membered heterocyclic boron compounds containing intramolecular N-B bond.
Solubility
Soluble in water 20°C 140g/L.
Notes
Store away from acid chloride, acid anhydrides and oxidizing agents. Keep the container tightly closed.
It is used as an intermediate in the preparation of 2.5-Dimethyl-2.5-bis(tertbutyl-peroxy)hexane to produce polyethylene copolymers and polyethylene rubbers. It is also used in the synthesis of six- and seven-membered heterocyclic boron compounds containing intramolecular N-B bond.
Solubility
Soluble in water 20°C 140g/L.
Notes
Store away from acid chloride, acid anhydrides and oxidizing agents. Keep the container tightly closed.
Specifications
Specifications
| Melting Point | 87°C to 91°C |
| Density | 0.898 |
| Boiling Point | 214°C to 215°C |
| Flash Point | 126°C (258°F) |
| Odor | Odorless |
| Quantity | 100 g |
| Beilstein | 1361437 |
| Solubility Information | Soluble in water 20°C 140g/L. |
| Formula Weight | 146.23 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | H318 |
| Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
| Precautionary Statement | P280-P305+P351+P338-P310 |
| EINECSNumber | 203-731-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only