missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-2,5-hexanediol, 97%

Catalog No. AAA1198436
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1198436 500 g
AAA1198422 100 g
AAA119840E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1198436 Supplier Thermo Scientific Chemicals Supplier No. A1198436
Only null left

CAS: 110-03-2 | C8H18O2 | 146.23 g/mol

It is used as an intermediate in the preparation of 2.5-Dimethyl-2.5-bis(tertbutyl-peroxy)hexane to produce polyethylene copolymers and polyethylene rubbers. It is also used in the synthesis of six- and seven-membered heterocyclic boron compounds containing intramolecular N-B bond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate in the preparation of 2.5-Dimethyl-2.5-bis(tertbutyl-peroxy)hexane to produce polyethylene copolymers and polyethylene rubbers. It is also used in the synthesis of six- and seven-membered heterocyclic boron compounds containing intramolecular N-B bond.

Solubility
Soluble in water 20°C 140g/L.

Notes
Store away from acid chloride, acid anhydrides and oxidizing agents. Keep the container tightly closed.

Chemical Identifiers

CAS 110-03-2
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00004473
InChI Key ZWNMRZQYWRLGMM-UHFFFAOYSA-N
Synonym 2,5-dimethyl-2,5-hexanediol, 2,5-hexanediol, 2,5-dimethyl, unii-3pnb2s8721, 1,1,4,4-tetramethyl-1,4-butanediol, 2,5-dihydroxy-2,5-dimethylhexane, 2,5-dimethyl-hexane-2,5-diol, dimethylhexanediol, dsstox_cid_6830, dsstox_rid_78222, dsstox_gsid_26830
PubChem CID 8031
IUPAC Name 2,5-dimethylhexane-2,5-diol
SMILES CC(C)(O)CCC(C)(C)O

Specifications

Melting Point 87°C to 91°C
Density 0.898
Boiling Point 214°C to 215°C
Flash Point 126°C (258°F)
Odor Odorless
Quantity 500 g
Beilstein 1361437
Solubility Information Soluble in water 20°C 140g/L.
Formula Weight 146.23
Percent Purity 97%
Chemical Name or Material 2,5-Dimethyl-2,5-hexanediol
Show More Show Less
Hazard Category H318
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 203-731-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only