missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-1,5-hexadien-3-ol, 97%

Catalog No. p-7024840
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2007206 5 g
AAB2007214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2007206 Supplier Thermo Scientific Chemicals Supplier No. B2007206
Only null left

CAS: 17123-63-6 | C8H14O | 126.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17123-63-6
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00026056
InChI Key QQMFDURNWCUKNS-UHFFFAOYSA-N
Synonym 2,5-dimethyl-1,5-hexadien-3-ol, acmc-1c55p, isopropenyl methallyl carbinol, 2,5-dimethyl-1,5-hexadiene-3-ol, 1,5-hexadien-3-ol,2,5-dimethyl
PubChem CID 551242
IUPAC Name 2,5-dimethylhexa-1,5-dien-3-ol
SMILES CC(=C)CC(C(=C)C)O

Specifications

Quantity 5 g
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material 2,5-Dimethyl-1,5-hexadien-3-ol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only