missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-1,3-oxazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog No. p-2697185
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC26801DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC26801DA Supplier Thermo Scientific Chemicals Supplier No. CC26801DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23000-14-8
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.126
MDL Number MFCD03011595
InChI Key LHGRUGVXZLHYKE-UHFFFAOYSA-N
Synonym 2,5-dimethyloxazole-4-carboxylic acid, dimethyl-1,3-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 2,5-dimethyl, 2,5-dimethyl-1,3-oxazole-4-carboxylicacid, 4-carboxy-2,5-dimethyl-1,3-oxazole, 2,5-dimethyl-oxazole-4-carboxylic acid, 2.5-dimethyl-1,3-oxazole-4-carboxylic acid
PubChem CID 2795465
IUPAC Name 2,5-dimethyl-1,3-oxazole-4-carboxylic acid
SMILES CC1=C(N=C(O1)C)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 141.13
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethyl-1, 3-oxazole-4-carboxylic acid
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.