missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethoxytetrahydrofuran (cis- and trans- mixture) 98.0+%, TCI America™
$82.02 - $635.65
Chemical Identifiers
| CAS | 696-59-3 |
|---|---|
| Molecular Formula | C6H12O3 |
| Molecular Weight (g/mol) | 132.159 |
| MDL Number | MFCD00005359 |
| InChI Key | GFISDBXSWQMOND-UHFFFAOYSA-N |
| Synonym | 2,5-dimethoxytetrahydrofuran, tetrahydro-2,5-dimethoxyfuran, furan, tetrahydro-2,5-dimethoxy, 2,5-dimethoxytetrahydrofurane, 2,5-dimethoxy-tetrahydrofuran, 2,5-dimethoxytetrahydrofuran, mixture of cis-and trans isomers, dimethoxytetrahydrofuran van, protectol dmt, furan,5-dimethoxy, dmthf |
| PubChem CID | 79098 |
| IUPAC Name | 2,5-dimethoxyoxolane |
| SMILES | COC1CCC(O1)OC |
Chemical Identifiers
| 696-59-3 | |
| 132.159 | |
| GFISDBXSWQMOND-UHFFFAOYSA-N | |
| 79098 | |
| COC1CCC(O1)OC |
| C6H12O3 | |
| MFCD00005359 | |
| 2,5-dimethoxytetrahydrofuran, tetrahydro-2,5-dimethoxyfuran, furan, tetrahydro-2,5-dimethoxy, 2,5-dimethoxytetrahydrofurane, 2,5-dimethoxy-tetrahydrofuran, 2,5-dimethoxytetrahydrofuran, mixture of cis-and trans isomers, dimethoxytetrahydrofuran van, protectol dmt, furan,5-dimethoxy, dmthf | |
| 2,5-dimethoxyoxolane |
Specifications
| 696-59-3 | |
| 146°C | |
| Clear Liquid at 20°C | |
| MFCD00005359 | |
| 2,5-dimethoxytetrahydrofuran, tetrahydro-2,5-dimethoxyfuran, furan, tetrahydro-2,5-dimethoxy, 2,5-dimethoxytetrahydrofurane, 2,5-dimethoxy-tetrahydrofuran, 2,5-dimethoxytetrahydrofuran, mixture of cis-and trans isomers, dimethoxytetrahydrofuran van, protectol dmt, furan,5-dimethoxy, dmthf | |
| COC1CCC(O1)OC | |
| 132.159 | |
| 132.16 | |
| 2,5-Dimethoxytetrahydrofuran (cis- and trans- mixture) |
| Yellow | |
| 25 g | |
| C6H12O3 | |
| 1993 | |
| GFISDBXSWQMOND-UHFFFAOYSA-N | |
| 2,5-dimethoxyoxolane | |
| 79098 | |
| ≥98.0% (GC) |
Safety and Handling
EINECSNumber : (5)-0054
TSCA : Yes