missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxytetrahydrofuran (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. D098625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D098625G 25 g
D0986100G 100 g
D0986500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D098625G Supplier TCI America Supplier No. D098625G

Chemical Identifiers

CAS 696-59-3
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00005359
InChI Key GFISDBXSWQMOND-UHFFFAOYSA-N
Synonym 2,5-dimethoxytetrahydrofuran, tetrahydro-2,5-dimethoxyfuran, furan, tetrahydro-2,5-dimethoxy, 2,5-dimethoxytetrahydrofurane, 2,5-dimethoxy-tetrahydrofuran, 2,5-dimethoxytetrahydrofuran, mixture of cis-and trans isomers, dimethoxytetrahydrofuran van, protectol dmt, furan,5-dimethoxy, dmthf
PubChem CID 79098
IUPAC Name 2,5-dimethoxyoxolane
SMILES COC1CCC(O1)OC

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
UN Number 1993
Boiling Point 146°C
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,5-Dimethoxytetrahydrofuran (cis- and trans- mixture)
EINECSNumber (5)-0054
TSCA Yes