missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxycinnamic Acid 97.0+%, TCI America™

Catalog No. D197210G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D197210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D197210G Supplier TCI America Supplier No. D197210G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10538-51-9
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00004378
InChI Key JPQWWJZORKTMIZ-ZZXKWVIFSA-N
Synonym 2,5-dimethoxycinnamic acid, 3-2,5-dimethoxyphenyl acrylic acid, e-3-2,5-dimethoxyphenyl acrylic acid, 2e-3-2,5-dimethoxyphenyl acrylic acid, 3-2,5-dimethoxyphenyl-2-propenoic acid, 2e-3-2,5-dimethoxyphenyl prop-2-enoic acid, 3-2,5-dimethoxyphenyl prop-2-enoic acid, rarechem bk hc t324, timtec-bb sbb000471, trans-2,5-dimethoxycinnamic acid
PubChem CID 697594
IUPAC Name (E)-3-(2,5-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC(=C(C=C1)OC)C=CC(=O)O

Specifications

Melting Point 148°C
Color Yellow
Quantity 10 g
Formula Weight 208.21
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethoxycinnamic Acid