missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxybenzaldehyde, 98+%

Catalog No. AAA1992836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1992836 500 g
AAA1992814 25 g
AAA1992822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1992836 Supplier Thermo Scientific Chemicals Supplier No. A1992836

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 93-02-7 | C9H10O3 | 166.176 g/mol

2,5-Dimethoxybenzaldehyde is used in the preparation of 2,5-dimethoxyphenethylamine, which is utilized to prepare psychoactive drugs such as 2,5-dimethoxy-4-bromophenethylamine, 2,5-dimethoxy-4-iodophenethylamine and 4-Chloro-2,5-dimethoxy-phenethylamine. It acts as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethoxybenzaldehyde is used in the preparation of 2,5-dimethoxyphenethylamine, which is utilized to prepare psychoactive drugs such as 2,5-dimethoxy-4-bromophenethylamine, 2,5-dimethoxy-4-iodophenethylamine and 4-Chloro-2,5-dimethoxy-phenethylamine. It acts as an intermediate in organic synthesis.

Solubility
Soluble in chloroform and methanol. Slightly soluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 93-02-7
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00003314
InChI Key AFUKNJHPZAVHGQ-UHFFFAOYSA-N
Synonym benzaldehyde, 2,5-dimethoxy, 2,5-dimethoxy benzaldehyde, unii-w49s1ppl78, 2,5-dimethoxybenzald, pubchem2176, gentisic aldehyde, 5-dimethoxy benzaldehyde, 2,5-dimethoxybenzaldehyd, 2,5-dimethoxybenzaldehye, 2.5-dimethoxybenzaldehyde
PubChem CID 66726
IUPAC Name 2,5-dimethoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)OC)C=O

Specifications

Melting Point 47°C to 51°C
Boiling Point 146°C (10 mmHg)
Flash Point >110°C (230°F)
Quantity 500 g
Beilstein 509301
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform and methanol. Slightly soluble in water.
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 2,5-Dimethoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P332+P313-P342+P311-P362-P501c
EINECSNumber 202-211-5
RTECSNumber CU5740500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only