missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxybenzaldehyde, 97%

Catalog No. p-4258964
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115390250 25 g Glass bottle
AC115391000 100 g Glass bottle
AC115395000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC115390250 Supplier Thermo Scientific Chemicals Supplier No. 115390250

CAS: 93-02-7 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93-02-7
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
InChI Key AFUKNJHPZAVHGQ-UHFFFAOYSA-N
Synonym benzaldehyde, 2,5-dimethoxy, 2,5-dimethoxy benzaldehyde, unii-w49s1ppl78, 2,5-dimethoxybenzald, pubchem2176, gentisic aldehyde, 5-dimethoxy benzaldehyde, 2,5-dimethoxybenzaldehyd, 2,5-dimethoxybenzaldehye, 2.5-dimethoxybenzaldehyde
PubChem CID 66726
IUPAC Name 2,5-dimethoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)OC)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 45.0°C to 50.0°C
Color Yellow to Beige
Boiling Point 146.0°C (10.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CHO
Quantity 25 g
Beilstein 08,245
Solubility Information Solubility in water: 795mg/l (25°C). Other solubilities: soluble in most common organic solvents
Formula Weight 166.18
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 2, 5-Dimethoxybenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 202-211-5

RUO – Research Use Only