missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethoxybenzaldehyde 97.0+%, TCI America™
$106.25 - $106.25
Chemical Identifiers
| CAS | 93-02-7 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.176 |
| MDL Number | MFCD00003314 |
| InChI Key | AFUKNJHPZAVHGQ-UHFFFAOYSA-N |
| Synonym | benzaldehyde, 2,5-dimethoxy, 2,5-dimethoxy benzaldehyde, unii-w49s1ppl78, 2,5-dimethoxybenzald, pubchem2176, gentisic aldehyde, 5-dimethoxy benzaldehyde, 2,5-dimethoxybenzaldehyd, 2,5-dimethoxybenzaldehye, 2.5-dimethoxybenzaldehyde |
| PubChem CID | 66726 |
| IUPAC Name | 2,5-dimethoxybenzaldehyde |
| SMILES | COC1=CC(=C(C=C1)OC)C=O |
Chemical Identifiers
| 93-02-7 | |
| 166.176 | |
| AFUKNJHPZAVHGQ-UHFFFAOYSA-N | |
| 66726 | |
| COC1=CC(=C(C=C1)OC)C=O |
| C9H10O3 | |
| MFCD00003314 | |
| benzaldehyde, 2,5-dimethoxy, 2,5-dimethoxy benzaldehyde, unii-w49s1ppl78, 2,5-dimethoxybenzald, pubchem2176, gentisic aldehyde, 5-dimethoxy benzaldehyde, 2,5-dimethoxybenzaldehyd, 2,5-dimethoxybenzaldehye, 2.5-dimethoxybenzaldehyde | |
| 2,5-dimethoxybenzaldehyde |
Specifications
| 93-02-7 | |
| 270°C | |
| MFCD00003314 | |
| benzaldehyde, 2,5-dimethoxy, 2,5-dimethoxy benzaldehyde, unii-w49s1ppl78, 2,5-dimethoxybenzald, pubchem2176, gentisic aldehyde, 5-dimethoxy benzaldehyde, 2,5-dimethoxybenzaldehyd, 2,5-dimethoxybenzaldehye, 2.5-dimethoxybenzaldehyde | |
| COC1=CC(=C(C=C1)OC)C=O | |
| 166.176 | |
| 166.18 | |
| Crystalline Powder |
| 50°C | |
| C9H10O3 | |
| 25 g | |
| AFUKNJHPZAVHGQ-UHFFFAOYSA-N | |
| 2,5-dimethoxybenzaldehyde | |
| 66726 | |
| ≥97.0% (GC) | |
| 2,5-Dimethoxybenzaldehyde |
Safety and Handling
EINECSNumber : (3)-2665
RTECSNumber : CU5740500
TSCA : Yes