missing translation for 'onlineSavingsMsg'
Learn More

2',5'-Dimethoxyacetophenone 98.0+%, TCI America™

Catalog No. p-7126731
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D144425G 25 g
D1444500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D144425G Supplier TCI America Supplier No. D144425G

Chemical Identifiers

CAS 1201-38-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00008728
InChI Key FAXUIYJKGGUCBO-UHFFFAOYSA-N
Synonym 2',5'-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethanone, 2,5-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethan-1-one, ethanone, 1-2,5-dimethoxyphenyl, 1-2,5-dimethoxyphenyl-ethanone, 2-acetyl-1,4-dimethoxybenzene, acetophenone, 2',5'-dimethoxy, 1-acetyl-2,5-dimethoxybenzene, pubchem13430
PubChem CID 70991
IUPAC Name 1-(2,5-dimethoxyphenyl)ethan-1-one
SMILES COC1=CC=C(OC)C(=C1)C(C)=O

Specifications

Color Yellow
Boiling Point 158°C
Quantity 25 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,5′-Dimethoxyacetophenone