missing translation for 'onlineSavingsMsg'
Learn More

2',5'-Dimethoxyacetophenone, 99%

CAS: 1201-38-3 | C10H12O3 | 180.20 g/mol

$155.62 - $155.62

Chemical Identifiers

CAS 1201-38-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00008728
InChI Key FAXUIYJKGGUCBO-UHFFFAOYSA-N
Synonym 2',5'-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethanone, 2,5-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethan-1-one, ethanone, 1-2,5-dimethoxyphenyl, 1-2,5-dimethoxyphenyl-ethanone, 2-acetyl-1,4-dimethoxybenzene, acetophenone, 2',5'-dimethoxy, 1-acetyl-2,5-dimethoxybenzene, pubchem13430
PubChem CID 70991
IUPAC Name 1-(2,5-dimethoxyphenyl)ethanone
SMILES COC1=CC=C(OC)C(=C1)C(C)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC115340250
SDS View Documents
Thermo Scientific Chemicals
115340250
25 g Glass bottle
Each for $155.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1201-38-3
180.20
FAXUIYJKGGUCBO-UHFFFAOYSA-N
70991
COC1=CC=C(OC)C(=C1)C(C)=O
C10H12O3
MFCD00008728
2',5'-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethanone, 2,5-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethan-1-one, ethanone, 1-2,5-dimethoxyphenyl, 1-2,5-dimethoxyphenyl-ethanone, 2-acetyl-1,4-dimethoxybenzene, acetophenone, 2',5'-dimethoxy, 1-acetyl-2,5-dimethoxybenzene, pubchem13430
1-(2,5-dimethoxyphenyl)ethanone

Specifications

1201-38-3
100.0
Green-Yellow to Yellow
155°C to 158°C (11.0 mmHg)
Authentic
Glass bottle
1.5421 to 1.5441
MFCD00008728
08, 271
2',5'-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethanone, 2,5-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethan-1-one, ethanone, 1-2,5-dimethoxyphenyl, 1-2,5-dimethoxyphenyl-ethanone, 2-acetyl-1,4-dimethoxybenzene, acetophenone, 2',5'-dimethoxy, 1-acetyl-2,5-dimethoxybenzene, pubchem13430
COC1=CC=C(OC)C(=C1)C(C)=O
180.20
180.2
Liquid
98.5
18°C to 20°C
1.1300g/mL
>112°C
98.5% min. (GC)
C10H12O3
(CH3O)2C6H3COCH3
25 g
1.13
FAXUIYJKGGUCBO-UHFFFAOYSA-N
1-(2,5-dimethoxyphenyl)ethanone
70991
99%
2', 5'-Dimethoxyacetophenone, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-858-0

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only