Learn More
2,5-Dimercapto-1,3,4-thiadiazole dipotassium salt, 97%
CAS: 4628-94-8 | C2K2N2S3 | 226.413 g/mol
Supplier: Thermo Scientific Chemicals L1110514
Description
1,3,4-thiadiazole-2,5-dithiol (Bismuthiol I) was used in determination of tellurium in indium phosphide doped with cadmium telluride by electro thermal atomic absorption spectrometry. It forms polymers with metals and can be useful in treatment of metal-polluted water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3,4-thiadiazole-2,5-dithiol (Bismuthiol I) was used in determination of tellurium in indium phosphide doped with cadmium telluride by electro thermal atomic absorption spectrometry. It forms polymers with metals and can be useful in treatment of metal-polluted water.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 2,5-Dimercapto-1,3,4-thiadiazole dipotassium salt | |
| ∽274°C (decomposition) | |
| MFCD00003104 | |
| 4917786 | |
| Insoluble in water. | |
| C1(=NN=C(S1)[S-])[S-].[K+].[K+] | |
| 226.413 | |
| 226.43 |
| 4628-94-8 | |
| C2K2N2S3 | |
| 25 g | |
| 2,5-dimercapto-1,3,4-thiadiazole, dipotassium salt, dipotassium 1,3,4-thiadiazole-2,5-dithiolate, dipotassium 1,3,4-thiadiazole-2,5-bis thiolate, 1,3,4-thiadiazolidine-2,5-dithione, dipotassium salt, 1,3,4-thiadiazolidine-2,5-dithione, potassium salt 1:2, dipotassium 5-sulfanidyl-1,3,4-thiadiazol-2-yl sulfanide, 2,5-dimercapto-1,3,4-thiadiazole dipotassium salt, 1,3,4-thiadiazole-2,5-dithiol dipotassium salt, 2,5-di potassiothio-1,3,4-thiadiazole | |
| GPWLFGDMYSVEGN-UHFFFAOYSA-L | |
| dipotassium;1,3,4-thiadiazole-2,5-dithiolate | |
| 78364 | |
| 97% |
Chemical Identifiers
| 4628-94-8 | |
| 226.413 | |
| GPWLFGDMYSVEGN-UHFFFAOYSA-L | |
| 78364 | |
| C1(=NN=C(S1)[S-])[S-].[K+].[K+] |
| C2K2N2S3 | |
| MFCD00003104 | |
| 2,5-dimercapto-1,3,4-thiadiazole, dipotassium salt, dipotassium 1,3,4-thiadiazole-2,5-dithiolate, dipotassium 1,3,4-thiadiazole-2,5-bis thiolate, 1,3,4-thiadiazolidine-2,5-dithione, dipotassium salt, 1,3,4-thiadiazolidine-2,5-dithione, potassium salt 1:2, dipotassium 5-sulfanidyl-1,3,4-thiadiazol-2-yl sulfanide, 2,5-dimercapto-1,3,4-thiadiazole dipotassium salt, 1,3,4-thiadiazole-2,5-dithiol dipotassium salt, 2,5-di potassiothio-1,3,4-thiadiazole | |
| dipotassium;1,3,4-thiadiazole-2,5-dithiolate |
Safety and Handling
EINECSNumber : 225-043-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only