missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimercapto-1,3,4-thiadiazole, 98%

Catalog No. p-113278
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115310250 25 g Plastic bottle
AC115311000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC115310250 Supplier Thermo Scientific Chemicals Supplier No. 115310250
Only null left

CAS: 1072-71-5 | C2H2N2S3 | 150.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1072-71-5
Molecular Formula C2H2N2S3
Molecular Weight (g/mol) 150.23
MDL Number MFCD00003103
InChI Key BIGYLAKFCGVRAN-UHFFFAOYSA-N
Synonym 1,3,4-thiadiazole-2,5-dithiol, 2,5-dimercapto-1,3,4-thiadiazole, bismuthiol i, dimercaptothiadiazole, bismuththiol, 2,5-dimercaptothiadiazole, usaf a-8354, bismuthiol, py 61h, bismuththiol i
PubChem CID 2723630
IUPAC Name 1,3,4-thiadiazolidine-2,5-dithione
SMILES S=C1NNC(=S)S1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 161°C to 165°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 25 g
Beilstein 27, 677
Solubility Information Solubility in water: 20g/L water (20°C)
Formula Weight 150.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,5-Dimercapto-1,3,4-thiadiazole, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-014-1
RTECSNumber XI3850000
TSCA TSCA

RUO – Research Use Only