missing translation for 'onlineSavingsMsg'
Learn More

2',5'-Dihydroxyacetophenone, 97%

Catalog No. AC114810050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114810050 5 g Glass bottle
AC114810250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC114810050 Supplier Thermo Scientific Chemicals Supplier No. 114810050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 490-78-8 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 490-78-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002343
InChI Key WLDWSGZHNBANIO-UHFFFAOYSA-N
Synonym 2',5'-dihydroxyacetophenone, 1-2,5-dihydroxyphenyl ethanone, 2-acetylhydroquinone, 2,5-dihydroxyacetophenone, quinacetophenone, acetylhydroquinone, ethanone, 1-2,5-dihydroxyphenyl, 1-2,5-dihydroxyphenyl ethan-1-one, acetylquinol, acetophenone, 2',5'-dihydroxy
PubChem CID 10279
IUPAC Name 1-(2,5-dihydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 203.0°C to 208.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (HO)2C6H3COCH3
Quantity 5 g
Beilstein 08, 271
Solubility Information Solubility in water: insoluble
Formula Weight 152.15
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2', 5'-Dihydroxyacetophenone, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-716-4
RTECSNumber AM7700000
TSCA TSCA

RUO – Research Use Only