missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxy-2,5-dimethyl-1,4-dithiane 95.0+%, TCI America™

Catalog No. p-7127913
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37855G 5 g
D378525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37855G Supplier TCI America Supplier No. D37855G
Only null left

Chemical Identifiers

CAS 55704-78-4
Molecular Formula C6H12O2S2
Molecular Weight (g/mol) 180.28
MDL Number MFCD00010121
InChI Key NHKIYYMFGJBOTK-UHFFFAOYSA-N
Synonym 2,5-dimethyl-2,5-dihydroxy-1,4-dithiane, dimeric mercapto propanone, 2-mercaptopropanone, 1,4-dithiane-2,5-diol, 2,5-dimethyl, 2,5-dimethyl-2,5-dihydroxy-p-dithiane, 2,5-dihydroxy-2,5-dimethyl-1,4-dithiane, 2,5-dimethyl-p-dithiane-2,5-diol, 2,5-dimethyl-1,4 dithiane-2,5-diol, fema no. 3450, dimericmercaptopropanone
PubChem CID 62105
IUPAC Name 2,5-dimethyl-1,4-dithiane-2,5-diol
SMILES CC1(CSC(CS1)(C)O)O

Specifications

Melting Point 88°C
Color White
Quantity 5 g
Formula Weight 180.28
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dihydroxy-2,5-dimethyl-1,4-dithiane