missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dihydrofuran 98.0+%, TCI America™
Supplier: TCI America D055125ML
Specifications
| 2,5-Dihydrofuran | |
| Yellow | |
| C4H6O | |
| 25 mL | |
| furan, 2,5-dihydro, 3-oxolene, 1-oxa-3-cyclopentene, 2,5-dihydro-furan, furan,5-dihydro, oxa-3-cyclopentene, 2,5 dihydrofurane, 2,5-dihydro-fura, pubchem3987, acmc-1buc6 | |
| C1C=CCO1 | |
| 70.091 | |
| 70.09 | |
| Liquid |
| 1708-29-8 | |
| 67°C | |
| MFCD00003219 | |
| 1993 | |
| ARGCQEVBJHPOGB-UHFFFAOYSA-N | |
| 2,5-dihydrofuran | |
| 15570 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1708-29-8 | |
| 70.091 | |
| ARGCQEVBJHPOGB-UHFFFAOYSA-N | |
| 15570 | |
| C1C=CCO1 |
| C4H6O | |
| MFCD00003219 | |
| furan, 2,5-dihydro, 3-oxolene, 1-oxa-3-cyclopentene, 2,5-dihydro-furan, furan,5-dihydro, oxa-3-cyclopentene, 2,5 dihydrofurane, 2,5-dihydro-fura, pubchem3987, acmc-1buc6 | |
| 2,5-dihydrofuran |
Safety and Handling
EINECSNumber : (9)-0731
RTECSNumber : LU0660000
TSCA : Yes