missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydrofuran 98.0+%, TCI America™

Catalog No. D0551500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0551500ML 500 mL
D055125ML 25 mL
D0551100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0551500ML Supplier TCI America Supplier No. D0551500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1708-29-8
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00003219
InChI Key ARGCQEVBJHPOGB-UHFFFAOYSA-N
Synonym furan, 2,5-dihydro, 3-oxolene, 1-oxa-3-cyclopentene, 2,5-dihydro-furan, furan,5-dihydro, oxa-3-cyclopentene, 2,5 dihydrofurane, 2,5-dihydro-fura, pubchem3987, acmc-1buc6
PubChem CID 15570
IUPAC Name 2,5-dihydrofuran
SMILES C1C=CCO1

Specifications

Color Yellow
Boiling Point 67°C
Quantity 500 mL
UN Number 1993
Formula Weight 70.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dihydrofuran
EINECSNumber (9)-0731
RTECSNumber LU0660000
TSCA Yes