missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoro-7,7,8,8-tetracyanoquinodimethane 98.0+%, TCI America™

Catalog No. d3200100mg
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
D3200100MG 100 mg
1 options

Catalog No. D3200100MG

Supplier: TCI America D3200100MG

Only null left

Chemical Identifiers

CAS 73318-02-2
Molecular Formula C12H2F2N4
Molecular Weight (g/mol) 240.173
InChI Key PFSDYUVXXQTNMX-UHFFFAOYSA-N
Synonym 2,5-TCNQF2
PubChem CID 5255840
IUPAC Name 2-[4-(dicyanomethylidene)-2,5-difluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile
SMILES C1=C(C(=C(C#N)C#N)C=C(C1=C(C#N)C#N)F)F

Specifications

Melting Point 270°C
Quantity 100 mg
UN Number 3439
Formula Weight 240.17
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Difluoro-7,7,8,8-tetracyanoquinodimethane