missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorothiophene-3-sulfonyl chloride, 97%

Catalog No. aal0646106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0646106 5 g
AAL0646103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0646106 Supplier Thermo Scientific Chemicals Supplier No. L0646106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56946-83-9 | C4HCl3O2S2 | 251.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56946-83-9
Molecular Formula C4HCl3O2S2
Molecular Weight (g/mol) 251.52
MDL Number MFCD00051665
InChI Key JJKSHSHZJOWSEC-UHFFFAOYSA-N
Synonym 2,5-dichlorothiophene-3-sulphonyl chloride, 2,5-dichloro-3-thiophenesulfonyl chloride, 3-thiophenesulfonyl chloride, 2,5-dichloro, 2,5-dichlorothiophene-3-sulfonic acid chloride, pubchem7412, 2,5-dichlorothiophene-3-sulfonylchloride, acmc-1avwk, 2,5-dichloro 3-thienyl chlorosulfone, 2,5-dichloro-3-thienylsulfonyl chloride, 2,5-dichlorothien-3-yl-sulfonyl chloride
PubChem CID 2736094
IUPAC Name 2,5-dichlorothiophene-3-sulfonyl chloride
SMILES C1=C(SC(=C1S(=O)(=O)Cl)Cl)Cl

Specifications

Density 1.697
Boiling Point 256°C to 257°C
Flash Point >110°C (230°F)
Refractive Index 1.597
Quantity 5 g
UN Number UN3265
Beilstein 1285315
Sensitivity Moisture sensitive
Formula Weight 251.54
Percent Purity 97%
Chemical Name or Material 2,5-Dichlorothiophene-3-sulfonyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only