missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloroterephthalonitrile 99.0+%, TCI America™

Catalog No. D17345G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17345G 5 g
D17341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17345G Supplier TCI America Supplier No. D17345G

Chemical Identifiers

CAS 1897-43-4
Molecular Formula C8H2Cl2N2
Molecular Weight (g/mol) 197.018
MDL Number MFCD00059584
InChI Key UCRQGBRIGDKUAM-UHFFFAOYSA-N
Synonym 1,4-Dichloro-2,5-dicyanobenzene
PubChem CID 121191
IUPAC Name 2,5-dichlorobenzene-1,4-dicarbonitrile
SMILES C1=C(C(=CC(=C1Cl)C#N)Cl)C#N

Specifications

Melting Point 214°C
Color White-Yellow
Quantity 5 g
Formula Weight 197.02
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloroterephthalonitrile
RTECSNumber CZ1939030
TSCA No