missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloropyrimidine 98.0+%, TCI America™

Catalog No. p-7128071
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40455G 5 g
D404525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40455G Supplier TCI America Supplier No. D40455G
Only null left

Chemical Identifiers

CAS 22536-67-0
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.974
MDL Number MFCD00673131
InChI Key CEJAHXLRNZJPQH-UHFFFAOYSA-N
Synonym 2,5-dichloro-pyrimidine, pyrimidine, 2,5-dichloro, 2,5-dichloro-1,3-diazine, pubchem13600, acmc-209fxc, ksc493a8n, 2,5-dichloropyrimidine, pyrimidine, 2,5-dichloro-7ci,8ci,9ci
PubChem CID 590641
IUPAC Name 2,5-dichloropyrimidine
SMILES C1=C(C=NC(=N1)Cl)Cl

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 190°C
Quantity 5 g
Formula Weight 148.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloropyrimidine