Learn More
2,5-Dichloropyrimidine, 95%
CAS: 22536-67-0 | C4H2Cl2N2 | 148.974 g/mol
Supplier: Thermo Scientific Chemicals H6406614
Description
2,5-Dichloropyrimidine acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,5-Dichloropyrimidine acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
| 2,5-Dichloropyrimidine | |
| 48°C to 55°C | |
| 107°C (225°F) | |
| MFCD00673131 | |
| UN3077 | |
| 2,5-dichloro-pyrimidine, pyrimidine, 2,5-dichloro, 2,5-dichloro-1,3-diazine, pubchem13600, acmc-209fxc, ksc493a8n, 2,5-dichloropyrimidine, pyrimidine, 2,5-dichloro-7ci,8ci,9ci | |
| CEJAHXLRNZJPQH-UHFFFAOYSA-N | |
| 2,5-dichloropyrimidine | |
| 590641 | |
| 95% |
| 22536-67-0 | |
| White | |
| C4H2Cl2N2 | |
| 25 g | |
| 110912 | |
| Slightly soluble in water. | |
| C1=C(C=NC(=N1)Cl)Cl | |
| 148.974 | |
| 148.97 | |
| Powder |
Chemical Identifiers
| 22536-67-0 | |
| 148.974 | |
| CEJAHXLRNZJPQH-UHFFFAOYSA-N | |
| 590641 | |
| C1=C(C=NC(=N1)Cl)Cl |
| C4H2Cl2N2 | |
| MFCD00673131 | |
| 2,5-dichloro-pyrimidine, pyrimidine, 2,5-dichloro, 2,5-dichloro-1,3-diazine, pubchem13600, acmc-209fxc, ksc493a8n, 2,5-dichloropyrimidine, pyrimidine, 2,5-dichloro-7ci,8ci,9ci | |
| 2,5-dichloropyrimidine |
Safety and Handling
GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only