missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloropyridine 99.0+%, TCI America™

Catalog No. D16075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16075G 5 g
D160725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16075G Supplier TCI America Supplier No. D16075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16110-09-1 | C5H3Cl2N | 147.99 g/mol

Chemical Identifiers

CAS 16110-09-1
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006239
InChI Key GCTFDMFLLBCLPF-UHFFFAOYSA-N
Synonym pyridine, 2,5-dichloro, 2,5-dichloro-pyridine, 2,5-dichloro pyridine, ccris 1718, 2,5-dichlorpyridin, 2,5-dichloropyridin, 2,5-dichioropyridine, 2.5-dichloropyridine, pubchem1199, 2,5 dichloropyridine
PubChem CID 27685
IUPAC Name 2,5-dichloropyridine
SMILES ClC1=CC=C(Cl)N=C1

Specifications

Melting Point 61°C
Color White
Boiling Point 98°C
Quantity 5 g
Formula Weight 147.99
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloropyridine
RTECSNumber US8225000
TSCA No