missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloropyridine-4-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7193926
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D50821G Supplier TCI America Supplier No. D50821G

Specifications

Chemical Name or Material 2,5-Dichloropyridine-4-boronic Acid (contains varying amounts of Anhydride)
CAS 847664-64-6
Melting Point 202°C
Color White
Molecular Formula C5H4BCl2NO2
Quantity 1 g
MDL Number MFCD06798254
Synonym 2,5-dichloropyridine-4-boronic acid, 2,5-dichloropyridin-4-yl-4-boronic acid, 2,5-dichloro-4-pyridyl boronic acid, 2,5-dichloropyridine-4-boronicacid, 2,5-dichloropyridin-4-yl boronic acid, 2,5-dichloro-4-pyridineboronic acid, boronic acid, b-2,5-dichloro-4-pyridinyl, boronic acid, 2,5-dichloro-4-pyridinyl, pubchem17071, acmc-209pwe
InChI Key BEJJMDOFMZCRST-UHFFFAOYSA-N
SMILES B(C1=CC(=NC=C1Cl)Cl)(O)O
IUPAC Name (2,5-dichloropyridin-4-yl)boronic acid
Molecular Weight (g/mol) 191.802
PubChem CID 12177416
Formula Weight 191.80
Physical Form Crystalline Powder
Show More Show Less