missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dichlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$175.20 - $175.20
Chemical Identifiers
| CAS | 135145-90-3 |
|---|---|
| Molecular Formula | C6H5BCl2O2 |
| Molecular Weight (g/mol) | 190.814 |
| MDL Number | MFCD01863182 |
| InChI Key | NNTFPBXQPOQRBT-UHFFFAOYSA-N |
| PubChem CID | 2773371 |
| IUPAC Name | (2,5-dichlorophenyl)boronic acid |
| SMILES | B(C1=C(C=CC(=C1)Cl)Cl)(O)O |
Chemical Identifiers
| 135145-90-3 | |
| 190.814 | |
| NNTFPBXQPOQRBT-UHFFFAOYSA-N | |
| (2,5-dichlorophenyl)boronic acid |
| C6H5BCl2O2 | |
| MFCD01863182 | |
| 2773371 | |
| B(C1=C(C=CC(=C1)Cl)Cl)(O)O |
Specifications
| 135145-90-3 | |
| C6H5BCl2O2 | |
| 5 g | |
| NNTFPBXQPOQRBT-UHFFFAOYSA-N | |
| (2,5-dichlorophenyl)boronic acid | |
| 2773371 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD01863182 | |
| 2,5-dichlorobenzeneboronic acid, 2,5-dichlorophenyl boronic acid, 2,5-dichlorophenylboronicacid, 2,5-dichlorophenyl boranediol, boronic acid, 2,5-dichlorophenyl, pubchem1811, acmc-209bxy, ksc174k3h, 2,5-dichloro-phenylboronic acid | |
| B(C1=C(C=CC(=C1)Cl)Cl)(O)O | |
| 190.814 | |
| 190.81 | |
| 2,5-Dichlorophenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA : No