missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D30445G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D30445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D30445G Supplier TCI America Supplier No. D30445G

Chemical Identifiers

CAS 135145-90-3
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.814
MDL Number MFCD01863182
InChI Key NNTFPBXQPOQRBT-UHFFFAOYSA-N
PubChem CID 2773371
IUPAC Name (2,5-dichlorophenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)Cl)Cl)(O)O

Specifications

Color White-Yellow
Quantity 5 g
Synonym 2,5-dichlorobenzeneboronic acid, 2,5-dichlorophenyl boronic acid, 2,5-dichlorophenylboronicacid, 2,5-dichlorophenyl boranediol, boronic acid, 2,5-dichlorophenyl, pubchem1811, acmc-209bxy, ksc174k3h, 2,5-dichloro-phenylboronic acid
Formula Weight 190.81
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichlorophenylboronic Acid (contains varying amounts of Anhydride)