missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorophenyl Isocyanate 97.0+%, TCI America™

Catalog No. D039725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D039725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D039725G Supplier TCI America Supplier No. D039725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5392-82-5
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.01
MDL Number MFCD00002002
InChI Key PEQMJVGRHNZPAM-UHFFFAOYSA-N
Synonym 2,5-dichlorophenyl isocyanate, benzene, 1,4-dichloro-2-isocyanato, 2,5-dichlorophenylisocyanate, isocyanic acid, 2,5-dichlorophenyl ester, isocyanic acid 2,5-dichlorophenyl ester, acmc-209laz, 2,5-dichlorobenzenisocyanate, benzene,4-dichloro-2-isocyanato, #, isocyanic acid,5-dichlorophenyl ester
PubChem CID 79360
IUPAC Name 1,4-dichloro-2-isocyanatobenzene
SMILES ClC1=CC=C(Cl)C(=C1)N=C=O

Specifications

Color White-Yellow
Boiling Point 126°C
Quantity 25 g
UN Number 2250
Formula Weight 188.01
Percent Purity ≥97.0% (GC)
Physical Form Crystals
Chemical Name or Material 2,5-Dichlorophenyl Isocyanate
TSCA Yes
Recommended Storage Refrigerator