missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloroaniline 98.0+%, TCI America™

Catalog No. p-7126143
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
D032225G 25 g
D0322100G 100 g
D0322500G 500 g
3 options

Catalog No. D032225G

Supplier: TCI America D032225G

Only null left

Chemical Identifiers

CAS 95-82-9
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.01
MDL Number MFCD00007667
InChI Key AVYGCQXNNJPXSS-UHFFFAOYSA-N
Synonym 1-amino-2,5-dichlorobenzene, azoic diazo component 3, scarlet 2g base, fast scarlet gg base, 2,5-dichloranilin, benzenamine, 2,5-dichloro, azobase dca, scarlet base gg, fast scarlet 2g, scarlet base ngg
PubChem CID 7262
ChEBI CHEBI:34245
IUPAC Name 2,5-dichloroaniline
SMILES NC1=CC(Cl)=CC=C1Cl

Specifications

Melting Point 52°C
Boiling Point 251°C
Quantity 25 g
UN Number 3442
Formula Weight 162.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloroaniline
EINECSNumber (3)-0261
RTECSNumber BX2610000
TSCA Yes