missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloroaniline 98.0+%, TCI America™

Catalog No. D032225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D032225G 25 g
D0322100G 100 g
D0322500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D032225G Supplier TCI America Supplier No. D032225G

Chemical Identifiers

CAS 95-82-9
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.01
MDL Number MFCD00007667
InChI Key AVYGCQXNNJPXSS-UHFFFAOYSA-N
Synonym 1-amino-2,5-dichlorobenzene, azoic diazo component 3, scarlet 2g base, fast scarlet gg base, 2,5-dichloranilin, benzenamine, 2,5-dichloro, azobase dca, scarlet base gg, fast scarlet 2g, scarlet base ngg
PubChem CID 7262
ChEBI CHEBI:34245
IUPAC Name 2,5-dichloroaniline
SMILES NC1=CC(Cl)=CC=C1Cl

Specifications

Melting Point 52°C
Boiling Point 251°C
Quantity 25 g
UN Number 3442
Formula Weight 162.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloroaniline
EINECSNumber (3)-0261
RTECSNumber BX2610000
TSCA Yes