missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-p-benzoquinone, 98%

Catalog No. p-7077267
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC406730050 5 g
AC406730250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC406730050 Supplier Thermo Scientific Chemicals Supplier No. 406730050

CAS: 615-93-0 | C6H2Cl2O2 | 176.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 615-93-0
Molecular Formula C6H2Cl2O2
Molecular Weight (g/mol) 176.99
InChI Key LNXVNZRYYHFMEY-UHFFFAOYSA-N
Synonym 2,5-dichloro-1,4-benzoquinone, 2,5-dichloro-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro, 2,5-dichlorobenzo-1,4-quinone, p-benzoquinone, 2,5-dichloro, 2,5-dichloro-p-quinone, acmc-1bhnt, 2,5-dichlorobenzoquinone, p-benzoquinone,5-dichloro, 2,5-dichoro-p-benzoquinone
PubChem CID 12011
IUPAC Name 2,5-dichlorocyclohexa-2,5-diene-1,4-dione
SMILES C1=C(C(=O)C=C(C1=O)Cl)Cl

Specifications

Melting Point 159.0°C to 161.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 176.99
Percent Purity 98%
Chemical Name or Material 2,5-Dichloro-p-benzoquinone
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 210-453-8

RUO – Research Use Only