missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-3,4-ethylenedioxythiophene 98.0+%, TCI America™

Catalog No. D47465G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47465G 5 g
D47461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47465G Supplier TCI America Supplier No. D47465G
Only null left

Chemical Identifiers

CAS 225518-49-0
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.056
MDL Number MFCD09907757
InChI Key UOJXEABHQWNIKN-UHFFFAOYSA-N
Synonym 5,7-Dichloro-2,3-dihydrothieno[3,4-b][1,4]dioxine
PubChem CID 22177487
IUPAC Name 5,7-dichloro-2,3-dihydrothieno[3,4-b][1,4]dioxine
SMILES C1COC2=C(SC(=C2O1)Cl)Cl

Specifications

Melting Point 67°C
Color White-Yellow
Quantity 5 g
Formula Weight 211.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloro-3,4-ethylenedioxythiophene
TSCA No
Recommended Storage Refrigerator