missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-1,4-benzoquinone 98.0+%, TCI America™

Catalog No. D03431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D03431G 1 g
D03435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D03431G Supplier TCI America Supplier No. D03431G

Chemical Identifiers

CAS 615-93-0
Molecular Formula C6H2Cl2O2
Molecular Weight (g/mol) 176.98
MDL Number MFCD00041736
InChI Key LNXVNZRYYHFMEY-UHFFFAOYSA-N
Synonym 2,5-dichloro-1,4-benzoquinone, 2,5-dichloro-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro, 2,5-dichlorobenzo-1,4-quinone, p-benzoquinone, 2,5-dichloro, 2,5-dichloro-p-quinone, acmc-1bhnt, 2,5-dichlorobenzoquinone, p-benzoquinone,5-dichloro, 2,5-dichoro-p-benzoquinone
PubChem CID 12011
IUPAC Name 2,5-dichlorocyclohexa-2,5-diene-1,4-dione
SMILES C1=C(C(=O)C=C(C1=O)Cl)Cl

Specifications

Melting Point 161°C
Color Yellow
Quantity 1 g
Formula Weight 176.98
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloro-1,4-benzoquinone
RTECSNumber DK3990000
TSCA Yes