missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromothiophene 95.0+%, TCI America™

Catalog No. p-7126784
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D154925G 25 g
D1549100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D154925G Supplier TCI America Supplier No. D154925G
Only null left

Chemical Identifiers

CAS 3141-27-3
Molecular Formula C4H2Br2S
Molecular Weight (g/mol) 241.93
MDL Number MFCD00005420
InChI Key KBVDUUXRXJTAJC-UHFFFAOYSA-N
Synonym thiophene, 2,5-dibromo, 2,5-dibromo-thiophene, unii-f0033k8cxm, pubchem5512, thiophene,5-dibromo, acmc-1ckmh, 2,5-bis bromanyl thiophene, ksc224g0j, 2,5-dibromothiophene, bidd:gt0086
PubChem CID 18453
IUPAC Name 2,5-dibromothiophene
SMILES BrC1=CC=C(Br)S1

Specifications

Boiling Point 210°C
Quantity 25 g
UN Number 2810
Formula Weight 241.93
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dibromothiophene