missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromothiophene, 95%

CAS: 3141-27-3 | C4H2Br2S | 241.93 g/mol

Supplier:  Thermo Scientific Chemicals A1340618

 View more versions of this product

Catalog No. AAA1340618


Only null left
Explore available promotions

Description

Description

2,5-Dibromothiophene, is mainly used as pharmaceutical intermediate and synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dibromothiophene, is mainly used as pharmaceutical intermediate and synthesis intermediate.

Solubility
It is soluble in chloroform and methanol. It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.Light Sensitive
Specifications

Specifications

2,5-Dibromothiophene
-6°C
210°C to 212°C
Pungent
1.629
50 g
109899
It is soluble in chloroform and methanol. It is insoluble in water.
BrC1=CC=C(Br)S1
241.93
241.94
3141-27-3
2.15
99°C (210°F)
C4H2Br2S
MFCD00005420
UN2810
thiophene, 2,5-dibromo, 2,5-dibromo-thiophene, unii-f0033k8cxm, pubchem5512, thiophene,5-dibromo, acmc-1ckmh, 2,5-bis bromanyl thiophene, ksc224g0j, 2,5-dibromothiophene, bidd:gt0086
KBVDUUXRXJTAJC-UHFFFAOYSA-N
2,5-dibromothiophene
18453
96%

Chemical Identifiers

3141-27-3
241.93
KBVDUUXRXJTAJC-UHFFFAOYSA-N
18453
BrC1=CC=C(Br)S1
C4H2Br2S
MFCD00005420
thiophene, 2,5-dibromo, 2,5-dibromo-thiophene, unii-f0033k8cxm, pubchem5512, thiophene,5-dibromo, acmc-1ckmh, 2,5-bis bromanyl thiophene, ksc224g0j, 2,5-dibromothiophene, bidd:gt0086
2,5-dibromothiophene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H341-H315-H319-H317-H335
Fatal if swallowed.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 221-547-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only