missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromothiophene, 95%

Catalog No. AAA1340614
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1340614 25 g
AAA1340618 50 g
AAA1340630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1340614 Supplier Thermo Scientific Chemicals Supplier No. A1340614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3141-27-3 | C4H2Br2S | 241.93 g/mol

2,5-Dibromothiophene, is mainly used as pharmaceutical intermediate and synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dibromothiophene, is mainly used as pharmaceutical intermediate and synthesis intermediate.

Solubility
It is soluble in chloroform and methanol. It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.Light Sensitive

Chemical Identifiers

CAS 3141-27-3
Molecular Formula C4H2Br2S
Molecular Weight (g/mol) 241.93
MDL Number MFCD00005420
InChI Key KBVDUUXRXJTAJC-UHFFFAOYSA-N
Synonym thiophene, 2,5-dibromo, 2,5-dibromo-thiophene, unii-f0033k8cxm, pubchem5512, thiophene,5-dibromo, acmc-1ckmh, 2,5-bis bromanyl thiophene, ksc224g0j, 2,5-dibromothiophene, bidd:gt0086
PubChem CID 18453
IUPAC Name 2,5-dibromothiophene
SMILES BrC1=CC=C(Br)S1

Specifications

Melting Point -6°C
Density 2.15
Boiling Point 210°C to 212°C
Flash Point 99°C (210°F)
Odor Pungent
Refractive Index 1.629
Quantity 25 g
UN Number UN2810
Beilstein 109899
Solubility Information It is soluble in chloroform and methanol. It is insoluble in water.
Formula Weight 241.94
Percent Purity 96%
Chemical Name or Material 2,5-Dibromothiophene
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H300-H341-H315-H319-H317-H335
Fatal if swallowed.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 221-547-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only