missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromothiophene-3-carboxylic Acid 96.0+%, TCI America™

Catalog No. p-7128351
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D45821G Supplier TCI America Supplier No. D45821G

Chemical Identifiers

CAS 7311-70-8
Molecular Formula C5H2Br2O2S
Molecular Weight (g/mol) 285.94
MDL Number MFCD08445656
InChI Key PZBUYFPAASJYSI-UHFFFAOYSA-N
Synonym 2,5-Dibromo-3-thenoic Acid
PubChem CID 268907
IUPAC Name 2,5-dibromothiophene-3-carboxylic acid
SMILES OC(=O)C1=C(Br)SC(Br)=C1

Specifications

Melting Point 179°C
Color White-Yellow
Quantity 1 g
Formula Weight 285.94
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromothiophene-3-carboxylic Acid