missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dibromoselenophene 98.0+%, TCI America™
$223.68 - $672.92
Chemical Identifiers
| CAS | 1755-36-8 |
|---|---|
| Molecular Formula | C4H2Br2Se |
| Molecular Weight (g/mol) | 288.839 |
| InChI Key | QAYFAXYTKFYUDZ-UHFFFAOYSA-N |
| PubChem CID | 13439265 |
| IUPAC Name | 2,5-dibromoselenophene |
| SMILES | C1=C([Se]C(=C1)Br)Br |
Chemical Identifiers
| 1755-36-8 | |
| 288.839 | |
| 13439265 | |
| C1=C([Se]C(=C1)Br)Br |
| C4H2Br2Se | |
| QAYFAXYTKFYUDZ-UHFFFAOYSA-N | |
| 2,5-dibromoselenophene |
Specifications
| 1755-36-8 | |
| 104°C | |
| 1 g | |
| QAYFAXYTKFYUDZ-UHFFFAOYSA-N | |
| 2,5-dibromoselenophene | |
| 13439265 | |
| ≥98.0% (GC) | |
| 2,5-Dibromoselenophene |
| Yellow | |
| C4H2Br2Se | |
| 3440 | |
| C1=C([Se]C(=C1)Br)Br | |
| 288.839 | |
| 288.83 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator