missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromopyridine, 97%

Catalog No. AC112900100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112900100 10 g Glass bottle
AC112900500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC112900100 Supplier Thermo Scientific Chemicals Supplier No. 112900100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 624-28-2 | C5H3Br2N | 236.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 624-28-2
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.89
MDL Number MFCD00006221
InChI Key ZHXUWDPHUQHFOV-UHFFFAOYSA-N
Synonym 2,5-dibromo pyridine, pyridine, 2,5-dibromo, 2,5-dibrompyridin, 2,5-dibromo-pyridine, 2,5-dibromopyridin, 3,6-dibromopyridine, 2,5,-dibromopyridine, 2,5-dibrompyridine, 2,5 dibromopyridine, pyridine,5-dibromo
PubChem CID 69353
IUPAC Name 2,5-dibromopyridine
SMILES BrC1=CC=C(Br)N=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 91.0°C to 96.0°C
Color Beige to Brown or White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C5H3N)Br2
Quantity 10 g
Beilstein 20,233
Solubility Information Solubility in water: insoluble
Formula Weight 236.89
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 5-Dibromopyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-839-6

RUO – Research Use Only