missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromopyridine, 97%

Catalog No. AAA1093709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1093709 10 g
AAA1093718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1093709 Supplier Thermo Scientific Chemicals Supplier No. A1093709
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 624-28-2 | C5H3Br2N | 236.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 624-28-2
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.89
MDL Number MFCD00006221
InChI Key ZHXUWDPHUQHFOV-UHFFFAOYSA-N
Synonym 2,5-dibromo pyridine, pyridine, 2,5-dibromo, 2,5-dibrompyridin, 2,5-dibromo-pyridine, 2,5-dibromopyridin, 3,6-dibromopyridine, 2,5,-dibromopyridine, 2,5-dibrompyridine, 2,5 dibromopyridine, pyridine,5-dibromo
PubChem CID 69353
IUPAC Name 2,5-dibromopyridine
SMILES BrC1=CC=C(Br)N=C1

Specifications

Melting Point 92°C to 96°C
Boiling Point 249°C
Quantity 10 g
Beilstein 109099
Formula Weight 236.9
Percent Purity 97%
Chemical Name or Material 2,5-Dibromopyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-839-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only