missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-N-n-octyl-3,4-thiophenedicarboximide 98.0+%, TCI America™

Catalog No. D4219200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4219200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4219200MG Supplier TCI America Supplier No. D4219200MG
Only null left

Chemical Identifiers

CAS 566939-58-0
Molecular Formula C14H17Br2NO2S
Molecular Weight (g/mol) 423.16
MDL Number MFCD18804055
InChI Key GSGMEQUXTCYOAU-UHFFFAOYSA-N
Synonym 1,3-Dibromo-5-n-octyl-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione
PubChem CID 66886624
IUPAC Name 1,3-dibromo-5-octyl-4H,5H,6H-thieno[3,4-c]pyrrole-4,6-dione
SMILES CCCCCCCCN1C(=O)C2=C(Br)SC(Br)=C2C1=O

Specifications

Melting Point 107°C
Quantity 200 mg
Formula Weight 423.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromo-N-n-octyl-3,4-thiophenedicarboximide