missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dibromo-4-methylpyridine 98.0+%, TCI America™
$50.33 - $128.62
Chemical Identifiers
| CAS | 3430-26-0 |
|---|---|
| Molecular Formula | C6H5Br2N |
| Molecular Weight (g/mol) | 250.921 |
| MDL Number | MFCD00234955 |
| InChI Key | WWJLJUAHQHXDGM-UHFFFAOYSA-N |
| PubChem CID | 2734429 |
| IUPAC Name | 2,5-dibromo-4-methylpyridine |
| SMILES | CC1=CC(=NC=C1Br)Br |
Chemical Identifiers
| 3430-26-0 | |
| 250.921 | |
| WWJLJUAHQHXDGM-UHFFFAOYSA-N | |
| 2,5-dibromo-4-methylpyridine |
| C6H5Br2N | |
| MFCD00234955 | |
| 2734429 | |
| CC1=CC(=NC=C1Br)Br |
Specifications
| 3430-26-0 | |
| Yellow | |
| MFCD00234955 | |
| 2,5-dibromo-4-picoline, pyridine, 2,5-dibromo-4-methyl, 2,5-dibromo-4-methyl-pyridine, pubchem2411, 2 pound not5-dibromo-4-methylpyridine, acmc-209i6i, 2, 5-dibromo-4-picoline, 2,5-dibromo-4-methypyridine, ksc495g9l, tpc-py104 | |
| CC1=CC(=NC=C1Br)Br | |
| 250.921 | |
| 250.92 | |
| Crystalline Powder |
| 40°C | |
| C6H5Br2N | |
| 1 g | |
| WWJLJUAHQHXDGM-UHFFFAOYSA-N | |
| 2,5-dibromo-4-methylpyridine | |
| 2734429 | |
| ≥98.0% (GC) | |
| 2,5-Dibromo-4-methylpyridine |
Safety and Handling
TSCA : No