missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-4-methylpyridine 98.0+%, TCI America™

Catalog No. D40581G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40581G 1 g
D40585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40581G Supplier TCI America Supplier No. D40581G

Chemical Identifiers

CAS 3430-26-0
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.921
MDL Number MFCD00234955
InChI Key WWJLJUAHQHXDGM-UHFFFAOYSA-N
PubChem CID 2734429
IUPAC Name 2,5-dibromo-4-methylpyridine
SMILES CC1=CC(=NC=C1Br)Br

Specifications

Melting Point 40°C
Color Yellow
Quantity 1 g
Synonym 2,5-dibromo-4-picoline, pyridine, 2,5-dibromo-4-methyl, 2,5-dibromo-4-methyl-pyridine, pubchem2411, 2 pound not5-dibromo-4-methylpyridine, acmc-209i6i, 2, 5-dibromo-4-picoline, 2,5-dibromo-4-methypyridine, ksc495g9l, tpc-py104
Formula Weight 250.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromo-4-methylpyridine