missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-4-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog No. AAH3045706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3045706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3045706 Supplier Thermo Scientific Chemicals Supplier No. H3045706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3430-26-0
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.921
MDL Number MFCD00234955
InChI Key WWJLJUAHQHXDGM-UHFFFAOYSA-N
Synonym 2,5-dibromo-4-picoline, pyridine, 2,5-dibromo-4-methyl, 2,5-dibromo-4-methyl-pyridine, pubchem2411, 2 pound not5-dibromo-4-methylpyridine, acmc-209i6i, 2, 5-dibromo-4-picoline, 2,5-dibromo-4-methypyridine, ksc495g9l, tpc-py104
PubChem CID 2734429
IUPAC Name 2,5-dibromo-4-methylpyridine
SMILES CC1=CC(=NC=C1Br)Br

Specifications

Melting Point 36°C to 40°C
Quantity 5 g
Formula Weight 250.92
Percent Purity 97%
Chemical Name or Material 2,5-Dibromo-4-methylpyridine
Hazard Category H302+H312+H332-H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only