missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-dodecylthiophene 97.0+%, TCI America™

Catalog No. D40315G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40315G Supplier TCI America Supplier No. D40315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 148256-63-7
Molecular Formula C16H26Br2S
Molecular Weight (g/mol) 410.252
MDL Number MFCD00274315
InChI Key ZIZGNWAMVWNAJT-UHFFFAOYSA-N
Synonym thiophene, 2,5-dibromo-3-dodecyl, 2,5-dibromo-3-laurylthiophene, 2,5-dibromo-3-dodecyl-thiophene, 2,5-bromo-3-dodecylthiophene, acmc-209cip, ksc496a6t, bidd:gt0375, thiophene,2,5-dibromo-3-dodecyl, 2,5-bis bromanyl-3-dodecyl-thiophene, 2,5-dibromo-3-dodecylthiophene
PubChem CID 5072754
IUPAC Name 2,5-dibromo-3-dodecylthiophene
SMILES CCCCCCCCCCCCC1=C(SC(=C1)Br)Br

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 g
Formula Weight 410.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dibromo-3-dodecylthiophene
TSCA No
Recommended Storage Refrigerator