missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-cyclohexylthiophene 97.0+%, TCI America™

Catalog No. D52471G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D52471G Supplier TCI America Supplier No. D52471G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 302912-44-3
Molecular Formula C10H12Br2S
Molecular Weight (g/mol) 324.074
MDL Number MFCD02683539
InChI Key GQDHHQWPBULMEB-UHFFFAOYSA-N
PubChem CID 3341004
IUPAC Name 2,5-dibromo-3-cyclohexylthiophene
SMILES C1CCC(CC1)C2=C(SC(=C2)Br)Br

Specifications

Color Yellow
Boiling Point 296°C
Quantity 1 g
Formula Weight 324.07
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dibromo-3-cyclohexylthiophene
TSCA No
Recommended Storage Refrigerator