missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3,4-thiophenedicarboxylic Anhydride 98.0+%, TCI America™

Catalog No. d4542200mg
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4542200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4542200MG Supplier TCI America Supplier No. D4542200MG
Only null left

Chemical Identifiers

CAS 1015423-45-6
Molecular Formula C6Br2O3S
Molecular Weight (g/mol) 311.93
MDL Number MFCD27923031
InChI Key OQMJQMGTDSIYNQ-UHFFFAOYSA-N
Synonym 4,6-Dibromothieno[3,4-c]furan-1,3-dione
PubChem CID 71721475
IUPAC Name dibromo-1H,3H-thieno[3,4-c]furan-1,3-dione
SMILES BrC1=C2C(=O)OC(=O)C2=C(Br)S1

Specifications

Quantity 200 mg
Formula Weight 311.93
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromo-3,4-thiophenedicarboxylic Anhydride