missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3,4-dinitrothiophene 98.0+%, TCI America™

Catalog No. D23715G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23715G 5 g
D23711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23715G Supplier TCI America Supplier No. D23715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52431-30-8
Molecular Formula C4Br2N2O4S
Molecular Weight (g/mol) 331.92
MDL Number MFCD00015537
InChI Key AHGHPBPARMANQD-UHFFFAOYSA-N
Synonym 2,5-dibromo-3,4-dinitro-thiophene, thiophene, 2,5-dibromo-3,4-dinitro, acmc-1apgk, ksc491i5j, 2,5-dibromo-3,4-dinitrothiophene gc
PubChem CID 257024
IUPAC Name 2,5-dibromo-3,4-dinitrothiophene
SMILES [O-][N+](=O)C1=C(Br)SC(Br)=C1[N+]([O-])=O

Specifications

Melting Point 137°C
Quantity 5 g
Formula Weight 331.92
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromo-3,4-dinitrothiophene