missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dibromo-1,4-benzoquinone 98.0+%, TCI America™
$456.70 - $1368.22
Chemical Identifiers
| CAS | 1633-14-3 |
|---|---|
| Molecular Formula | C6H2Br2O2 |
| Molecular Weight (g/mol) | 265.888 |
| MDL Number | MFCD00156859 |
| InChI Key | KNPAQJBQOIAPBP-UHFFFAOYSA-N |
| Synonym | 2,5-Dibromo-p-quinone |
| PubChem CID | 543093 |
| IUPAC Name | 2,5-dibromocyclohexa-2,5-diene-1,4-dione |
| SMILES | C1=C(C(=O)C=C(C1=O)Br)Br |
Chemical Identifiers
| 1633-14-3 | |
| 265.888 | |
| KNPAQJBQOIAPBP-UHFFFAOYSA-N | |
| 543093 | |
| C1=C(C(=O)C=C(C1=O)Br)Br |
| C6H2Br2O2 | |
| MFCD00156859 | |
| 2,5-Dibromo-p-quinone | |
| 2,5-dibromocyclohexa-2,5-diene-1,4-dione |
Specifications
| 1633-14-3 | |
| C6H2Br2O2 | |
| 1 g | |
| KNPAQJBQOIAPBP-UHFFFAOYSA-N | |
| 2,5-dibromocyclohexa-2,5-diene-1,4-dione | |
| 543093 | |
| ≥98.0% (GC) | |
| 2,5-Dibromo-1,4-benzoquinone |
| 193°C | |
| MFCD00156859 | |
| 2,5-Dibromo-p-quinone | |
| C1=C(C(=O)C=C(C1=O)Br)Br | |
| 265.888 | |
| 265.89 | |
| Crystalline Powder |
Safety and Handling
TSCA : No