missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-1,4-benzoquinone 98.0+%, TCI America™

Catalog No. D22491G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22491G 1 g
D22495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22491G Supplier TCI America Supplier No. D22491G
Only null left

Chemical Identifiers

CAS 1633-14-3
Molecular Formula C6H2Br2O2
Molecular Weight (g/mol) 265.888
MDL Number MFCD00156859
InChI Key KNPAQJBQOIAPBP-UHFFFAOYSA-N
Synonym 2,5-Dibromo-p-quinone
PubChem CID 543093
IUPAC Name 2,5-dibromocyclohexa-2,5-diene-1,4-dione
SMILES C1=C(C(=O)C=C(C1=O)Br)Br

Specifications

Melting Point 193°C
Quantity 1 g
Formula Weight 265.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromo-1,4-benzoquinone